Assay Details
Target Gene Details
Entrez Gene ID: | 23560 |
Gene Name: | GTP binding protein 4 |
Gene Aliases: |
CRFG, NGB, NOG1 |
Location: |
Chr.10:988409-1017768 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP4 | NM_012341.2 | NP_036473.2 | ||
| AF120334.1 | AAD09830.1 | |||
| AF325353.1 | AAK13444.1 | |||
| AK001548.1 | ||||
| AK001552.1 | BAA91752.1 | |||
| AK097093.1 | ||||
| AK222861.1 | BAD96581.1 | |||
| AK295231.1 | ||||
| AK301721.1 | ||||
| AK302219.1 | ||||
| BC033784.1 | AAH33784.1 | |||
| BC038975.2 | AAH38975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
| nsv1161921 | Chr.10:823011 - 1033401 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map