Assay Details
Target Gene Details
Entrez Gene ID: | 23555 |
Gene Name: | tetraspanin 15 |
Gene Aliases: |
2700063A19Rik, NET-7, NET7, TM4SF15 |
Location: |
Chr.10:69451464-69549508 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN15 | NM_012339.3 | NP_036471.1 | ||
| XM_005269667.3 | XP_005269724.1 | |||
| XM_017016010.1 | XP_016871499.1 | |||
| AF120266.1 | AAD17295.1 | |||
| BC003157.1 | AAH03157.1 | |||
| BC004161.2 | AAH04161.1 |
More Information
Set Membership: |
|

Genomic Map