Assay Details
Target Gene Details
Entrez Gene ID: | 9126 |
Gene Name: | structural maintenance of chromosomes 3 |
Gene Aliases: |
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1 |
Location: |
Chr.10:110567691-110604634 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC3 | NM_005445.3 | NP_005436.1 | ||
| AF020043.1 | AAC14893.1 | |||
| AK289771.1 | ||||
| AK315845.1 | ||||
| BC047324.1 | AAH47324.1 | |||
| CA439554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526452 | Chr.10:110503272 - 110659020 on Build GRCh38 | Gain |
|
| nsv1054449 | Chr.10:110521913 - 110599006 on Build GRCh38 | Gain |
|
| nsv508607 | Chr.10:110571773 - 110655342 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map