Assay Details
Target Gene Details
Entrez Gene ID: | 143279 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 2 |
Gene Aliases: |
- |
Location: |
Chr.10:91409280-91514829 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECTD2 | NM_001284274.1 | NP_001271203.1 | ||
| NM_173497.3 | NP_775768.3 | |||
| NM_182765.4 | NP_877497.3 | |||
| NR_104291.1 | ||||
| XM_017015750.1 | XP_016871239.1 | |||
| XM_017015751.1 | XP_016871240.1 | |||
| AK094625.1 | BAC04388.1 | |||
| AK295723.1 | ||||
| BC040187.1 | AAH40187.1 | |||
| BX537856.1 | ||||
| HY158536.1 |
Target Gene Details
Entrez Gene ID: | 100188947 |
Gene Name: | HECTD2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:91306962-91611460 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECTD2-AS1 | NR_024467.1 | |||
| DA898704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040403 | Chr.10:91406062 - 91465279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map