Assay Details
Target Gene Details
Entrez Gene ID: | 100505933 |
Gene Name: | ADD3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:109945559-110008381 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADD3-AS1 | NR_038943.1 | |||
| AI425042.1 | ||||
| BU561061.1 | ||||
| BX537640.1 | ||||
| U92992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052255 | Chr.10:109950120 - 110033444 on Build GRCh38 | Gain |
|

Genomic Map