Assay Details
Target Gene Details
Entrez Gene ID: | 103344931 |
Gene Name: | uncharacterized LOC103344931 |
Gene Aliases: |
- |
Location: |
Chr.10:112823496-112827732 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC103344931 | NR_120684.1 | 1 | 1131 | |
| AL049387.1 | 1 | 261 |
Target Gene Details
Entrez Gene ID: | 143187 |
Gene Name: | vesicle transport through interaction with t-SNAREs 1A |
Gene Aliases: |
MMDS3, MVti1, VTI1RP2, Vti1-rp2 |
Location: |
Chr.10:112446988-112947204 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VTI1A | XM_005269546.3 | 9 | 10315 | XP_005269603.1 |
| XM_006717637.2 | XP_006717700.1 | |||
| XM_011539331.2 | XP_011537633.1 | |||
| XM_017015747.1 | XP_016871236.1 | |||
| XM_017015748.1 | XP_016871237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv285e212 | Chr.10:112807440 - 112830969 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map