Assay Details
Target Gene Details
Entrez Gene ID: | 23522 |
Gene Name: | lysine acetyltransferase 6B |
Gene Aliases: |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
Location: |
Chr.10:74824927-75032623 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6B | XM_005269664.2 | XP_005269721.1 | ||
| XM_017016000.1 | XP_016871489.1 | |||
| XM_017016001.1 | XP_016871490.1 | |||
| XM_017016003.1 | XP_016871492.1 | |||
| XM_017016004.1 | XP_016871493.1 | |||
| XM_017016005.1 | XP_016871494.1 | |||
| XM_017016006.1 | XP_016871495.1 | |||
| XM_017016007.1 | XP_016871496.1 | |||
| XM_017016008.1 | XP_016871497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551535 | Chr.10:74745813 - 74842047 on Build GRCh38 | Loss |
|
| esv2674729 | Chr.10:74824387 - 74831334 on Build GRCh38 | Deletion |
|
| nsv551536 | Chr.10:74825347 - 74827100 on Build GRCh38 | Loss |
|
| esv21440 | Chr.10:74825162 - 74827957 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map