Assay Details
Target Gene Details
Entrez Gene ID: | 102723166 |
Gene Name: | STAM antisense RNA 1 (head to head) |
Gene Aliases: |
locus3182 |
Location: |
Chr.10:17641284-17643918 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAM-AS1 | NR_110370.1 | 1 | 2084 | |
| AK123677.1 | 1 | 2045 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622495 | Chr.10:17638251 - 17642955 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112589999] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map