Assay Details
Target Gene Details
Entrez Gene ID: | 120 |
Gene Name: | adducin 3 |
Gene Aliases: |
ADDL, CPSQ3 |
Location: |
Chr.10:109996350-110135565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADD3 | NM_001121.3 | NP_001112.2 | ||
| NM_001320591.1 | NP_001307520.1 | |||
| NM_001320592.1 | NP_001307521.1 | |||
| NM_001320593.1 | NP_001307522.1 | |||
| NM_001320594.1 | NP_001307523.1 | |||
| NM_016824.4 | NP_058432.1 | |||
| NM_019903.4 | NP_063968.1 | |||
| AB209810.1 | BAD93047.1 | |||
| AK223270.1 | BAD96990.1 | |||
| AK291488.1 | ||||
| AL600737.1 | ||||
| BC062559.1 | AAH62559.1 | |||
| BI544596.1 | ||||
| BX647403.1 | CAI46048.1 | |||
| BX648016.1 | ||||
| CN428885.1 | ||||
| D67031.1 | BAA23783.1 | |||
| DA072039.1 | ||||
| DA477660.1 | ||||
| DA700119.1 | ||||
| EF553523.1 | ||||
| U37122.1 | AAB17126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7547 | Chr.10:109978521 - 110023780 on Build GRCh38 | Deletion |
|
| nsv1052255 | Chr.10:109950120 - 110033444 on Build GRCh38 | Gain |
|
| nsv951845 | Chr.10:110008343 - 110084542 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map