Assay Details
Target Gene Details
Entrez Gene ID: | 132 |
Gene Name: | adenosine kinase |
Gene Aliases: |
AK |
Location: |
Chr.10:74151185-74709303 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADK | NM_001123.3 | NP_001114.2 | ||
| NM_001202449.1 | NP_001189378.1 | |||
| NM_001202450.1 | NP_001189379.1 | |||
| NM_006721.3 | NP_006712.2 | |||
| XM_017015698.1 | XP_016871187.1 | |||
| XM_017015700.1 | XP_016871189.1 | |||
| XM_017015701.1 | XP_016871190.1 | |||
| XM_017015702.1 | XP_016871191.1 | |||
| XM_017015704.1 | XP_016871193.1 | |||
| XM_017015705.1 | XP_016871194.1 | |||
| XM_017015706.1 | XP_016871195.1 | |||
| AK290633.1 | ||||
| AK301590.1 | ||||
| AK302706.1 | ||||
| BC003568.1 | AAH03568.1 | |||
| BM472149.1 | ||||
| CB133126.1 | ||||
| DA356691.1 | ||||
| U33936.1 | AAB01689.1 | |||
| U50196.1 | AAA97893.1 | |||
| U90338.1 | AAB50234.1 | |||
| U90339.1 | AAB50235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2659855 | Chr.10:74162507 - 74240402 on Build GRCh38 | Deletion |
|
| esv3623825 | Chr.10:74175523 - 74208165 on Build GRCh38 | Loss |
|
| nsv515824 | Chr.10:74163688 - 74699911 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80017012] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map