Assay Details
Target Gene Details
Entrez Gene ID: | 54838 |
Gene Name: | WW domain binding protein 1-like |
Gene Aliases: |
C10orf26, OPA1L, OPAL1 |
Location: |
Chr.10:102742708-102816267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WBP1L | NM_001083913.1 | NP_001077382.1 | ||
| XM_017016360.1 | XP_016871849.1 | |||
| AK300871.1 | ||||
| AK308843.1 | ||||
| AK316452.1 | ||||
| CX163170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831969 | Chr.10:102651251 - 102812041 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74702335] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map