Assay Details
Target Gene Details
Entrez Gene ID: | 9585 |
Gene Name: | kinesin family member 20B |
Gene Aliases: |
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1 |
Location: |
Chr.10:89701590-89774943 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF20B | NM_001284259.1 | NP_001271188.1 | ||
| NM_016195.3 | NP_057279.2 | |||
| AB033337.1 | BAB69456.1 | |||
| AK074336.1 | ||||
| AK290439.1 | ||||
| AL117496.1 | CAB55962.1 | |||
| AY282406.1 | AAP40330.1 | |||
| BC046134.1 | AAH46134.1 | |||
| BC058913.1 | AAH58913.1 | |||
| BC093089.1 | AAH93089.1 | |||
| BC108688.1 | AAI08689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054572 | Chr.10:89712199 - 89777313 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map