Assay Details
Target Gene Details
Entrez Gene ID: | 22931 |
Gene Name: | RAB18, member RAS oncogene family |
Gene Aliases: |
RAB18LI1, WARBM3 |
Location: |
Chr.10:27504174-27542237 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB18 | NM_001256410.1 | NP_001243339.1 | ||
| NM_001256411.1 | NP_001243340.1 | |||
| NM_001256412.1 | NP_001243341.1 | |||
| NM_001256415.1 | NP_001243344.1 | |||
| NM_021252.4 | NP_067075.1 | |||
| NR_046172.1 | ||||
| AF087860.1 | AAP97170.1 | |||
| AF136974.1 | AAG49435.1 | |||
| AF137372.1 | AAF61433.1 | |||
| AF498950.1 | AAM21098.1 | |||
| AK001555.1 | ||||
| AK223153.1 | BAD96873.1 | |||
| AK295443.1 | ||||
| AK297629.1 | ||||
| AK299457.1 | ||||
| AL136734.1 | CAB66668.1 | |||
| AY574034.1 | AAU08232.1 | |||
| BC015014.2 | AAH15014.1 | |||
| BC029350.1 | AAH29350.1 | |||
| BP210053.1 | ||||
| BT009840.1 | AAP88842.1 | |||
| CN357871.1 | ||||
| CR533455.1 | CAG38486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045257 | Chr.10:27283511 - 27638017 on Build GRCh38 | Gain |
|
| nsv550241 | Chr.10:27280592 - 27736842 on Build GRCh38 | Gain |
|
| esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
|
| nsv8614 | Chr.10:27215558 - 27539477 on Build GRCh38 | Loss |
|
| esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map