Assay Details
Target Gene Details
Entrez Gene ID: | 22986 |
Gene Name: | sortilin related VPS10 domain containing receptor 3 |
Gene Aliases: |
SORCS |
Location: |
Chr.10:104641101-105265242 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS3 | NM_014978.2 | NP_055793.1 | ||
| AB028982.1 | BAA83011.2 | |||
| BC117417.1 |
Target Gene Details
Entrez Gene ID: | 100505890 |
Gene Name: | SORCS3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:104664608-104666216 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS3-AS1 | NR_104028.1 | 1 | 690 | |
| AK097854.1 | 1 | 690 | ||
| BU852305.1 | 1 | 693 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579100 | Chr.10:104573121 - 104983037 on Build GRCh38 | Loss |
|
| nsv831972 | Chr.10:104523290 - 104689830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map