Assay Details
Target Gene Details
Entrez Gene ID: | 9148 |
Gene Name: | neuralized E3 ubiquitin protein ligase 1 |
Gene Aliases: |
NEUR1, NEURL, RNF67, bA416N2.1, neu, neu-1 |
Location: |
Chr.10:103493890-103592552 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL1 | NM_004210.4 | NP_004201.3 | ||
| XM_011540331.2 | XP_011538633.1 | |||
| XM_011540335.2 | XP_011538637.1 | |||
| AF029729.1 | AAD01887.1 | |||
| BC026336.1 | AAH26336.1 | |||
| U87864.1 | AAC17474.1 |
Target Gene Details
Entrez Gene ID: | 102724341 |
Gene Name: | NEURL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:103479603-103517442 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL1-AS1 | NR_120675.1 | |||
| AA399019.1 | ||||
| BX089018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524966 | Chr.10:103441820 - 103717405 on Build GRCh38 | Loss |
|
| dgv966n100 | Chr.10:103385173 - 103553473 on Build GRCh38 | Gain |
|
| esv3624417 | Chr.10:103504194 - 103506754 on Build GRCh38 | Loss |
|
| esv3577694 | Chr.10:103391276 - 103553473 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map