Assay Details
Target Gene Details
Entrez Gene ID: | 142910 |
Gene Name: | lipase family member J |
Gene Aliases: |
LIPL1, bA425M17.2 |
Location: |
Chr.10:88583914-88623161 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPJ | XM_017015740.1 | XP_016871229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551839 | Chr.10:88575078 - 88662379 on Build GRCh38 | Loss |
|
| nsv1037657 | Chr.10:88570153 - 88608133 on Build GRCh38 | Loss |
|
| esv3624132 | Chr.10:88489037 - 88688695 on Build GRCh38 | Gain |
|
| esv3624134 | Chr.10:88555882 - 88703649 on Build GRCh38 | Gain |
|
| esv3624135 | Chr.10:88594312 - 88611587 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map