Assay Details
Target Gene Details
Entrez Gene ID: | 84991 |
Gene Name: | RNA binding motif protein 17 |
Gene Aliases: |
SPF45 |
Location: |
Chr.10:6088986-6117459 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM17 | NM_001145547.1 | NP_001139019.1 | ||
| NM_032905.4 | NP_116294.1 | |||
| AF542550.1 | AAQ09533.1 | |||
| AK314952.1 | ||||
| BC007871.2 | AAH07871.1 | |||
| BC039322.1 | AAH39322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040397 | Chr.10:6087796 - 6116734 on Build GRCh38 | Loss |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map