Assay Details
Target Gene Details
Entrez Gene ID: | 5995 |
Gene Name: | retinal G protein coupled receptor |
Gene Aliases: |
RP44 |
Location: |
Chr.10:84245053-84259188 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGR | NM_001012720.1 | NP_001012738.1 | ||
| NM_001012722.1 | NP_001012740.1 | |||
| NM_002921.3 | NP_002912.2 | |||
| BC008094.1 | AAH08094.1 | |||
| BC011349.1 | AAH11349.1 | |||
| BG912392.1 | ||||
| BX641033.1 | 2 | 856 | ||
| KU178307.1 | ||||
| KU178308.1 | ||||
| U14910.1 | AAA56748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain |
|
| esv3624030 | Chr.10:84248220 - 84249061 on Build GRCh38 | Loss |
|
| nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map