Assay Details
Target Gene Details
Entrez Gene ID: | 1355 |
Gene Name: | COX15, cytochrome c oxidase assembly homolog |
Gene Aliases: |
CEMCOX2 |
Location: |
Chr.10:99694793-99732667 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX15 | NM_001320974.1 | NP_001307903.1 | ||
| NM_001320975.1 | NP_001307904.1 | |||
| NM_001320976.1 | NP_001307905.1 | |||
| NM_004376.6 | NP_004367.2 | |||
| NM_078470.5 | NP_510870.1 | |||
| XM_006717634.3 | XP_006717697.1 | |||
| XM_011539298.2 | XP_011537600.1 | |||
| XM_017015715.1 | XP_016871204.1 | |||
| AF026850.1 | AAD08639.1 | |||
| AF044323.1 | AAD08646.1 | |||
| AK291654.1 | ||||
| AK298864.1 | ||||
| BC002382.2 | AAH02382.3 | |||
| BC013403.2 | AAH13403.1 | |||
| BC078161.1 | AAH78161.1 | |||
| BT007129.1 | AAP35793.1 | |||
| BX537557.1 | CAD97781.1 |
More Information
Set Membership: |
|

Genomic Map