Assay Details
Target Gene Details
Entrez Gene ID: | 102723166 |
Gene Name: | STAM antisense RNA 1 (head to head) |
Gene Aliases: |
locus3182 |
Location: |
Chr.10:17641284-17643918 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAM-AS1 | NR_110370.1 | 1 | 378 | |
| AK123677.1 | 1 | 339 |

Genomic Map