Assay Details
Target Gene Details
Entrez Gene ID: | 1979 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 2 |
Gene Aliases: |
4EBP2, PHASII |
Location: |
Chr.10:70404105-70428618 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4EBP2 | NM_004096.4 | NP_004087.1 | ||
| AK313724.1 | ||||
| BC005057.1 | AAH05057.1 | |||
| BC050633.1 | AAH50633.1 | |||
| BG717474.1 | ||||
| BT007317.1 | AAP35981.1 | |||
| CR542240.1 | CAG47036.1 | |||
| L36056.1 | AAA62270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map