Assay Details
Target Gene Details
Entrez Gene ID: | 104326055 |
Gene Name: | APOA1 antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.11:116836117-116855729 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOA1-AS | NR_126362.1 | |||
| DA327409.1 |
Target Gene Details
Entrez Gene ID: | 23387 |
Gene Name: | SIK family kinase 3 |
Gene Aliases: |
L19, QSK, SIK-3 |
Location: |
Chr.11:116843402-117098453 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627836 | Chr.11:116853047 - 116856370 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map