Assay Details
Target Gene Details
Entrez Gene ID: | 254187 |
Gene Name: | testis specific 10 interacting protein |
Gene Aliases: |
FAM161C |
Location: |
Chr.11:65945445-65960437 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSGA10IP | NM_152762.2 | NP_689975.2 | ||
| XM_011544887.2 | XP_011543189.1 | |||
| XM_011544888.2 | XP_011543190.1 | |||
| AK057442.1 | BAB71488.1 | |||
| BC104022.1 | AAI04023.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983036 | Chr.11:65954093 - 65958375 on Build GRCh38 | Duplication |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| esv2667131 | Chr.11:65955937 - 65960615 on Build GRCh38 | Deletion |
|
| esv3626685 | Chr.11:65955847 - 65960643 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72926898] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map