Assay Details
Target Gene Details
Entrez Gene ID: | 84623 |
Gene Name: | kin of IRRE like 3 (Drosophila) |
Gene Aliases: |
KIRRE, MRD4, NEPH2, PRO4502 |
Location: |
Chr.11:126423493-127003460 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIRREL3 | NM_001161707.1 | NP_001155179.1 | ||
| NM_001301097.1 | NP_001288026.1 | |||
| NM_032531.3 | NP_115920.1 | |||
| XM_011543026.2 | XP_011541328.1 | |||
| XM_011543027.2 | XP_011541329.1 | |||
| XM_011543028.2 | XP_011541330.1 | |||
| XM_011543030.2 | XP_011541332.1 | |||
| XM_011543031.2 | XP_011541333.1 | |||
| XM_011543032.2 | XP_011541334.1 | |||
| XM_011543033.1 | XP_011541335.1 | |||
| XM_017018418.1 | XP_016873907.1 | |||
| XM_017018419.1 | XP_016873908.1 | |||
| XM_017018420.1 | XP_016873909.1 | |||
| AB058770.1 | ||||
| AF480410.1 | AAN73042.1 | |||
| AK092047.1 | ||||
| AY358743.1 | AAQ89103.1 | |||
| BC101775.1 | AAI01776.1 | |||
| BC101801.1 | AAI01802.1 |
Target Gene Details
Entrez Gene ID: | 101929427 |
Gene Name: | uncharacterized LOC101929427 |
Gene Aliases: |
- |
Location: |
Chr.11:126652778-126682104 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929427 | NR_120532.1 | |||
| AA573248.1 |
More Information
Set Membership: |
|

Genomic Map