Assay Details
Target Gene Details
Entrez Gene ID: | 406 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like |
Gene Aliases: |
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5 |
Location: |
Chr.11:13276613-13387268 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map