Assay Details
Target Gene Details
Entrez Gene ID: | 1207 |
Gene Name: | chloride nucleotide-sensitive channel 1A |
Gene Aliases: |
CLCI, CLNS1B, ICln |
Location: |
Chr.11:77616151-77637806 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLNS1A | NM_001293.2 | NP_001284.1 | ||
| NM_001311199.1 | NP_001298128.1 | |||
| NM_001311200.1 | NP_001298129.1 | |||
| NM_001311201.1 | NP_001298130.1 | |||
| NM_001311202.1 | NP_001298131.1 | |||
| AF005422.1 | AAB61444.1 | |||
| AF026003.1 | AAB88806.1 | |||
| AK315259.1 | ||||
| AL530926.3 | ||||
| BC119634.1 | ||||
| BC119635.1 | ||||
| BQ643786.1 | ||||
| BT019907.1 | AAV38710.1 | |||
| BU169662.1 | ||||
| BU538016.1 | ||||
| U17899.1 | AAC50111.1 | |||
| U53454.1 | AAB03316.1 | |||
| X91788.1 | CAA62902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1234n100 | Chr.11:77572991 - 77724786 on Build GRCh38 | Gain |
|
| nsv555452 | Chr.11:77590296 - 77709269 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115786252] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map