Assay Details
Target Gene Details
Entrez Gene ID: | 27315 |
Gene Name: | post-GPI attachment to proteins 2 |
Gene Aliases: |
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21 |
Location: |
Chr.11:3797724-3826371 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAP2 | NM_001145438.2 | NP_001138910.1 | ||
| NM_001256235.1 | NP_001243164.1 | |||
| NM_001256236.1 | NP_001243165.1 | |||
| NM_001256237.1 | NP_001243166.1 | |||
| NM_001283038.1 | NP_001269967.1 | |||
| NM_001283039.1 | NP_001269968.1 | |||
| NM_001283040.1 | NP_001269969.1 | |||
| NR_045923.1 | ||||
| NR_045925.1 | ||||
| NR_045926.1 | ||||
| NR_045927.1 | ||||
| NR_104270.1 | ||||
| NR_104271.1 | ||||
| XM_006718185.2 | XP_006718248.1 | |||
| XM_011520000.1 | XP_011518302.1 | |||
| AL096753.1 | CAB46428.3 | |||
| DA287291.1 |
More Information
Set Membership: |
|

Genomic Map