Assay Details
Target Gene Details
Entrez Gene ID: | 5029 |
Gene Name: | purinergic receptor P2Y2 |
Gene Aliases: |
HP2U, P2RU1, P2U, P2U1, P2UR, P2Y2, P2Y2R |
Location: |
Chr.11:73200416-73246743 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY2 | NM_002564.3 | NP_002555.3 | ||
| NM_176071.2 | NP_788085.2 | |||
| NM_176072.2 | NP_788086.2 | |||
| XM_005274019.4 | XP_005274076.1 | |||
| XM_005274020.4 | 1 | 764 | XP_005274077.1 | |
| XM_005274021.4 | XP_005274078.1 | |||
| XM_011545074.2 | XP_011543376.1 | |||
| XM_017017839.1 | XP_016873328.1 | |||
| AK313942.1 | ||||
| BC012104.1 | AAH12104.1 | |||
| BC028135.1 | AAH28135.1 | |||
| U07225.1 | AAC04923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038032 | Chr.11:73214302 - 73255397 on Build GRCh38 | Gain |
|
| nsv832208 | Chr.11:73165139 - 73379958 on Build GRCh38 | Gain |
|
| esv3626812 | Chr.11:73213163 - 73237183 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map