Assay Details
Target Gene Details
Entrez Gene ID: | 11145 |
Gene Name: | phospholipase A2 group XVI |
Gene Aliases: |
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3 |
Location: |
Chr.11:63574462-63616926 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G16 | NM_001128203.1 | NP_001121675.1 | ||
| NM_007069.3 | NP_009000.2 | |||
| XM_006718426.1 | XP_006718489.1 | |||
| XM_011544741.1 | XP_011543043.1 | |||
| AB030814.1 | BAB08108.1 | |||
| AB439591.1 | ||||
| AF317086.1 | AAL26892.1 | |||
| AK313075.1 | ||||
| AX775893.1 | CAE11646.1 | |||
| BC001387.2 | AAH01387.1 | |||
| BC103807.2 | AAI03808.1 | |||
| BC103808.2 | AAI03809.1 | |||
| BG719903.1 | ||||
| CK003948.1 | ||||
| X92814.1 | CAA63423.1 |
More Information
Set Membership: |
|

Genomic Map