Assay Details
Target Gene Details
Entrez Gene ID: | 25988 |
Gene Name: | histone H4 transcription factor |
Gene Aliases: |
HiNF-P, MIZF, ZNF743 |
Location: |
Chr.11:119121523-119135055 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv514 | Chr.11:119114552 - 119143217 on Build GRCh38 | Deletion |
|
| nsv556466 | Chr.11:119106009 - 119188695 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map