Assay Details
Target Gene Details
Entrez Gene ID: | 406 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like |
Gene Aliases: |
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5 |
Location: |
Chr.11:13276613-13387268 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL | XM_011520105.2 | 1 | 249 | XP_011518407.2 |
| XM_011520107.2 | 1 | 244 | XP_011518409.2 | |
| XM_017017738.1 | 1 | 247 | XP_016873227.1 | |
| XM_017017739.1 | 1 | 243 | XP_016873228.1 |
More Information
Set Membership: |
|

Genomic Map