Assay Details
Target Gene Details
Entrez Gene ID: | 4076 |
Gene Name: | cell cycle associated protein 1 |
Gene Aliases: |
GPIAP1, GPIP137, GRIP137, M11S1, RNG105, p137GPI |
Location: |
Chr.11:34051617-34102610 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPRIN1 | NM_005898.4 | NP_005889.3 | ||
| NM_203364.2 | NP_976240.1 | |||
| XM_017017749.1 | XP_016873238.1 | |||
| AK127972.1 | ||||
| AK299720.1 | ||||
| BC001731.1 | AAH01731.2 | |||
| Z48042.1 | CAA88096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625906 | Chr.11:34068151 - 34093059 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map