Assay Details
Target Gene Details
Entrez Gene ID: | 63876 |
Gene Name: | PBX/knotted 1 homeobox 2 |
Gene Aliases: |
PREP2 |
Location: |
Chr.11:125111997-125433389 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKNOX2 | XM_017018110.1 | XP_016873599.1 |
Target Gene Details
Entrez Gene ID: | 103695364 |
Gene Name: | PKNOX2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.11:125158461-125164609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKNOX2-AS1 | NR_125759.1 | |||
| BP872841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556488 | Chr.11:125111809 - 125296916 on Build GRCh38 | Gain |
|
| nsv832290 | Chr.11:125112706 - 125271154 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map