Assay Details
Target Gene Details
Entrez Gene ID: | 55693 |
Gene Name: | lysine demethylase 4D |
Gene Aliases: |
JMJD2D |
Location: |
Chr.11:94973681-94999514 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4D | NM_018039.2 | NP_060509.2 | ||
| AW593120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv446 | Chr.11:94960555 - 95005748 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map