Assay Details
Target Gene Details
Entrez Gene ID: | 6157 |
Gene Name: | ribosomal protein L27a |
Gene Aliases: |
L27A |
Location: |
Chr.11:8682448-8689872 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL27A | NM_000990.4 | 5 | 3528 | NP_000981.1 |
| AK125453.1 | 1 | 1477 |
More Information
Set Membership: |
|

Genomic Map