Assay Details
Target Gene Details
Entrez Gene ID: | 9645 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 2 |
Gene Aliases: |
MICAL-2, MICAL2PV1, MICAL2PV2 |
Location: |
Chr.11:12110576-12281017 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICAL2 | NM_001282663.1 | NP_001269592.1 | ||
| NM_001282668.1 | NP_001269597.1 | |||
| NM_014632.3 | NP_055447.1 | |||
| AB018293.1 | ||||
| AK302580.1 | ||||
| AK302893.1 | ||||
| BC044577.1 | AAH44577.1 | |||
| DA246062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv5666 | Chr.11:12130749 - 12131595 on Build GRCh38 | Loss |
|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| nsv832068 | Chr.11:12098790 - 12284080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map