Assay Details
Target Gene Details
Entrez Gene ID: | 8898 |
Gene Name: | myotubularin related protein 2 |
Gene Aliases: |
CMT4B, CMT4B1 |
Location: |
Chr.11:95832880-95924207 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR2 | NM_001243571.1 | NP_001230500.1 | ||
| NM_016156.5 | NP_057240.3 | |||
| NM_201278.2 | NP_958435.1 | |||
| NM_201281.2 | NP_958438.1 | |||
| XM_005274374.2 | XP_005274431.1 | |||
| XM_005274375.2 | XP_005274432.1 | |||
| XM_006718934.2 | XP_006718997.1 | |||
| XM_006718935.2 | XP_006718998.1 | |||
| XM_006718936.3 | XP_006718999.1 | |||
| XM_011543058.2 | XP_011541360.1 | |||
| XM_011543059.2 | XP_011541361.1 | |||
| XM_017018517.1 | XP_016874006.1 | |||
| XM_017018518.1 | XP_016874007.1 | |||
| AB028996.1 | ||||
| AK027038.1 | ||||
| AK291277.1 | ||||
| AK293243.1 | ||||
| AK302940.1 | ||||
| BC052990.1 | AAH52990.1 | |||
| BX647997.1 | ||||
| U58033.1 | AAC79118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832235 | Chr.11:95753612 - 95935071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map