Assay Details
Target Gene Details
Entrez Gene ID: | 440028 |
Gene Name: | uncharacterized LOC440028 |
Gene Aliases: |
- |
Location: |
Chr.11:9754770-9759533 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC440028 | NR_033972.1 | 3 | 1376 | |
| AK090613.1 | 3 | 1376 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744095 | Chr.11:9623376 - 10133785 on Build GRCh38 | Deletion |
|
| nsv1045550 | Chr.11:9696777 - 9768541 on Build GRCh38 | Gain |
|
| nsv1053369 | Chr.11:9626290 - 9854507 on Build GRCh38 | Gain |
|
| nsv553468 | Chr.11:9646281 - 9806324 on Build GRCh38 | Gain |
|

Genomic Map