Assay Details
Target Gene Details
Entrez Gene ID: | 120425 |
Gene Name: | junction adhesion molecule like |
Gene Aliases: |
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638 |
Location: |
Chr.11:118193727-118225094 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAML | NM_001098526.1 | NP_001091996.1 | ||
| NM_001286570.1 | NP_001273499.1 | |||
| XM_017017213.1 | XP_016872702.1 | |||
| XM_017017214.1 | XP_016872703.1 | |||
| XM_017017215.1 | XP_016872704.1 | |||
| AJ515553.2 | CAD56620.2 | |||
| AK090409.1 | BAC03390.1 | |||
| AK298722.1 | ||||
| AK300851.1 | ||||
| AK301099.1 | ||||
| AY093686.1 | AAM15730.1 | |||
| AY138965.1 | AAN52117.1 | |||
| AY358362.1 | AAQ88728.1 | |||
| DC296115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511 | Chr.11:118204437 - 118239426 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75990945] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map