Assay Details
Target Gene Details
Entrez Gene ID: | 120071 |
Gene Name: | LARGE xylosyl- and glucuronyltransferase 2 |
Gene Aliases: |
GYLTL1B, PP5656 |
Location: |
Chr.11:45921621-45929096 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARGE2 | NM_001300722.1 | NP_001287651.1 | ||
| NM_152312.4 | NP_689525.3 | |||
| AK055829.1 | ||||
| AK096021.1 | BAC04675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516442 | Chr.11:45878411 - 45961834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map