Assay Details
Target Gene Details
Entrez Gene ID: | 53942 |
Gene Name: | contactin 5 |
Gene Aliases: |
HNB-2s, NB-2 |
Location: |
Chr.11:99020904-100358885 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTN5 | NM_001243270.1 | NP_001230199.1 | ||
| NM_001243271.1 | NP_001230200.1 | |||
| NM_014361.3 | NP_055176.1 | |||
| XM_011542871.1 | XP_011541173.1 | |||
| XM_011542873.1 | XP_011541175.1 | |||
| XM_017017926.1 | XP_016873415.1 | |||
| XM_017017927.1 | XP_016873416.1 | |||
| XM_017017928.1 | XP_016873417.1 | |||
| XM_017017929.1 | XP_016873418.1 | |||
| AK308788.1 | ||||
| BC039255.1 | AAH39255.1 | |||
| BC130619.1 | ||||
| BC144185.1 | ||||
| BC144186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428265 | Chr.11:98936705 - 99229707 on Build GRCh38 | Loss |
|
| nsv556046 | Chr.11:99024489 - 99058829 on Build GRCh38 | Loss |
|
| nsv832242 | Chr.11:98893493 - 99087654 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map