Assay Details
Target Gene Details
Entrez Gene ID: | 51642 |
Gene Name: | mitochondrial ribosomal protein L48 |
Gene Aliases: |
CGI-118, HSPC290, L48MT, MRP-L48 |
Location: |
Chr.11:73787872-73864611 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPL48 | NM_001318498.1 | NP_001305427.1 | ||
NM_001318499.1 | NP_001305428.1 | |||
NM_001318500.1 | NP_001305429.1 | |||
NM_016055.5 | NP_057139.1 | |||
NR_134658.1 | ||||
NR_134659.1 | ||||
NR_134660.1 | ||||
NR_134661.1 | ||||
XM_017017919.1 | XP_016873408.1 | |||
AF151876.1 | AAD34113.1 | |||
AF161408.1 | AAF28968.1 | |||
AK297746.1 | ||||
AK309219.1 | ||||
AK310005.1 | ||||
BC009764.1 | AAH09764.1 | |||
BI549412.1 | ||||
BM817807.1 | ||||
BM828996.1 | ||||
BP210564.1 | ||||
BU600573.1 | ||||
BU932890.1 | ||||
BX354271.2 | ||||
CF541319.1 | ||||
DA549653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509430 | Chr.11:73794575 - 73860940 on Build GRCh38 | Insertion |
![]() |
nsv391 | Chr.11:73797814 - 73832998 on Build GRCh38 | Insertion |
![]() |
nsv1045149 | Chr.11:73603520 - 73870863 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
