Assay Details
Target Gene Details
Entrez Gene ID: | 4322 |
Gene Name: | matrix metallopeptidase 13 |
Gene Aliases: |
CLG3, MANDP1, MMP-13 |
Location: |
Chr.11:102942992-102955734 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP13 | NM_002427.3 | NP_002418.1 | ||
| AK222748.1 | BAD96468.1 | |||
| AK292211.1 | ||||
| AK315341.1 | ||||
| AY256443.1 | AAP78939.1 | |||
| BC067522.1 | AAH67522.1 | |||
| BC067523.1 | AAH67523.1 | |||
| BC074807.2 | AAH74807.1 | |||
| BC074808.2 | AAH74808.1 | |||
| X75308.1 | CAA53056.1 | |||
| X81334.1 | CAA57108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464 | Chr.11:102922044 - 102954570 on Build GRCh38 | Insertion |
|
| nsv465 | Chr.11:102926638 - 102971487 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map