Assay Details
Target Gene Details
Entrez Gene ID: | 60488 |
Gene Name: | mitochondrial ribosomal protein S35 |
Gene Aliases: |
HDCMD11P, MDS023, MRP-S28, MRPS28 |
Location: |
Chr.12:27710773-27756304 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS35 | NM_001190864.1 | NP_001177793.1 | ||
| NM_021821.3 | NP_068593.2 | |||
| XM_017019780.1 | XP_016875269.1 | |||
| AF182422.1 | AAG14958.1 | |||
| AK075378.1 | BAC11579.1 | |||
| AK075515.1 | BAC11664.1 | |||
| AL512733.1 | CAC21665.1 | |||
| BC015862.1 | ||||
| BC028346.2 | AAH28346.2 | |||
| BC063515.1 | AAH63515.1 | |||
| BC109372.1 | AAI09373.1 | |||
| DA747094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| nsv557823 | Chr.12:27695850 - 27762012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map