Assay Details
Target Gene Details
Entrez Gene ID: | 81539 |
Gene Name: | solute carrier family 38 member 1 |
Gene Aliases: |
ATA1, NAT2, SAT1, SNAT1 |
Location: |
Chr.12:46183055-46269425 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A1 | NM_001077484.1 | 17 | 4739 | NP_001070952.1 |
| NM_001278387.1 | 18 | 5201 | NP_001265316.1 | |
| NM_001278388.1 | 16 | 4963 | NP_001265317.1 | |
| NM_001278389.1 | 17 | 4228 | NP_001265318.1 | |
| NM_030674.3 | 17 | 5078 | NP_109599.3 | |
| AK024263.1 | 1 | 678 | ||
| AK025900.1 | ||||
| AK056670.1 | 1 | 1372 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826360 | Chr.12:46121591 - 46204460 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map