Assay Details
Target Gene Details
Entrez Gene ID: | 29953 |
Gene Name: | thyrotropin releasing hormone degrading enzyme |
Gene Aliases: |
PAP-II, PGPEP2, TRH-DE |
Location: |
Chr.12:72087288-72666662 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRHDE | XM_017019244.1 | XP_016874733.1 |
Target Gene Details
Entrez Gene ID: | 283392 |
Gene Name: | TRHDE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:72253507-72273509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRHDE-AS1 | NR_026836.1 | |||
| NR_026837.1 | ||||
| AK095156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832452 | Chr.12:72191191 - 72370715 on Build GRCh38 | Gain |
|
| nsv1044083 | Chr.12:72253860 - 72881746 on Build GRCh38 | Gain |
|
| nsv559362 | Chr.12:72248004 - 72300911 on Build GRCh38 | Loss |
|
| esv2422452 | Chr.12:72092153 - 72781801 on Build GRCh38 | Duplication |
|
| nsv559363 | Chr.12:72262145 - 72865334 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map