Assay Details
Target Gene Details
Entrez Gene ID: | 219402 |
Gene Name: | mitochondrial translational initiation factor 3 |
Gene Aliases: |
IF3mt |
Location: |
Chr.13:27435639-27450900 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTIF3 | NM_001166261.1 | NP_001159733.1 | ||
| NM_001166262.1 | NP_001159734.1 | |||
| NM_001166263.1 | NP_001159735.1 | |||
| NM_152912.4 | NP_690876.3 | |||
| XM_006719771.3 | XP_006719834.1 | |||
| XM_006719772.3 | XP_006719835.1 | |||
| XM_011534957.2 | XP_011533259.1 | |||
| XM_011534958.2 | XP_011533260.1 | |||
| XM_011534959.2 | XP_011533261.1 | |||
| XM_011534960.2 | XP_011533262.1 | |||
| XM_011534961.2 | XP_011533263.1 | |||
| XM_011534962.2 | XP_011533264.1 | |||
| XM_011534963.2 | XP_011533265.1 | |||
| XM_017020417.1 | XP_016875906.1 | |||
| AF410851.1 | AAL04150.1 | |||
| AK094420.1 | ||||
| BC039599.1 | ||||
| BC046166.1 | AAH46166.1 | |||
| BG617536.1 | ||||
| BI460170.1 | ||||
| BM548061.1 | ||||
| BM768315.1 | ||||
| CN272769.1 |
More Information
Set Membership: |
|

Genomic Map