Assay Details
Target Gene Details
Entrez Gene ID: | 241 |
Gene Name: | arachidonate 5-lipoxygenase activating protein |
Gene Aliases: |
FLAP |
Location: |
Chr.13:30713478-30764428 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX5AP | NM_001204406.1 | NP_001191335.1 | ||
| AK314143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832574 | Chr.13:30657196 - 30822408 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114641250] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map