Assay Details
Target Gene Details
Entrez Gene ID: | 54841 |
Gene Name: | basic, immunoglobulin-like variable motif containing |
Gene Aliases: |
- |
Location: |
Chr.13:102799049-102841538 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIVM | NM_001159596.1 | NP_001153068.1 | ||
| NM_017693.3 | NP_060163.2 | |||
| AF339783.1 | ||||
| AF339793.1 | ||||
| AF411385.1 | AAM45761.1 | |||
| AK000166.1 | BAA90986.1 | |||
| AK225229.1 | ||||
| AK290658.1 | ||||
| AK304391.1 | ||||
| BC051813.2 | AAH51813.2 | |||
| BC075084.2 | AAH75084.1 | |||
| BC075085.2 | AAH75085.1 | |||
| BC112339.1 | AAI12340.1 |
Target Gene Details
Entrez Gene ID: | 100533467 |
Gene Name: | BIVM-ERCC5 readthrough |
Gene Aliases: |
ERCC5-202 |
Location: |
Chr.13:102807146-102876001 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIVM-ERCC5 | NM_001204425.1 | NP_001191354.1 | ||
| AB209312.1 | BAD92549.1 | |||
| LM994822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562981 | Chr.13:102820720 - 103931125 on Build GRCh38 | Gain |
|
| nsv1048497 | Chr.13:102768928 - 102842048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map