Assay Details
Target Gene Details
Entrez Gene ID: | 79587 |
Gene Name: | cysteinyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
COXPD27, cysRS |
Location: |
Chr.13:110641410-110713527 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARS2 | NM_024537.3 | NP_078813.1 | ||
| XM_006719953.2 | XP_006720016.1 | |||
| XM_011521116.1 | XP_011519418.1 | |||
| XM_011521117.2 | XP_011519419.1 | |||
| XM_011521118.2 | XP_011519420.1 | |||
| XM_017020741.1 | XP_016876230.1 | |||
| XM_017020742.1 | XP_016876231.1 | |||
| AB062436.1 | BAB93499.1 | |||
| AK022180.1 | BAB13978.1 | |||
| AK301907.1 | ||||
| BC007220.1 | AAH07220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain |
|
| nsv1042232 | Chr.13:110629776 - 110696128 on Build GRCh38 | Gain |
|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141446308] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map